Learn More
Scientific Industries Inc BALANCE PRECISION 120 GRAM DFS Item
Supplier:  Scientific Industries Inc AD120
Precision Scale with 120g capacity, resolution of 1mg, rugged die-cast metal housing and highest quality electromagnetic load-cell. Loaded with 7 weighing functions all essential units of measure, equipped with an RS232, USB, Clock, Backlit LCD Display.
Catalog No. 501139346
Only null left
Explore available promotions