Learn More
Scientific Industries Inc BALANCE PHARMACY CLASS II DFS Item
Supplier:  Scientific Industries Inc DRX4120
Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing. Capacity 120g x 0.001g, with a tare capacity equal to its range, and provides a display of scale capacity used (in percent) whenever weighing.
Catalog No. 501139343
Only null left
Explore available promotions