Learn More
Fujifilm Irvine Scientific BalanCD HEK293 | Powder | 10l
Catalog No. NC1845790
Encompass
BalanCD HEK293 medium is a versatile, chemically-defined, animal component-free medium optimized for transfection and production of viral vectors and recombinant proteins.
Catalog No. NC1845790 Supplier Fujifilm Irvine Scientific Supplier No. 9413710L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BalanCD HEK293 medium is a versatile, chemically-defined, animal component-free medium optimized for transfection and production of viral vectors and recombinant proteins.