Learn More
Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
Supplier: Bal Supply 20165WS
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
- 05-885C and 05-885E through K have screw-locking device
- Work well on both O-ring and ball-and-socket joints
- Others are intended for ball-and-socket joints only
Specifications
| 6/Pk. | |
| Stainless Steel | |
| 65 |
| Pinch Clamp | |
| Screw Locking Device |