Learn More
Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
Supplier:  Bal Supply 20135WS
 View more versions of this product
Catalog No. 05885G
Only null left
Description
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
  • 05-885C and 05-885E through K have screw-locking device
  • Work well on both O-ring and ball-and-socket joints
  • Others are intended for ball-and-socket joints only
Specifications
Specifications
12/Pk.
Stainless Steel
35
Pinch Clamp
Screw Locking Device
Product Suggestions
Product Suggestions