Learn More
Avantor™ BAKERBOND Speedisk™ SAX Quaternary Amine Extraction Disk
Description
- Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
- Require minimal glassware, set up time, and assembly
- Minimizes impact of extractable compounds
- Resists clogging and ensures high throughput rate
- Reduce SPE run times
Specifications
Specifications
| For Use With (Application) | EPA Method 552.1, Haloacetic Acids and Dalapon |
| For Use With (Equipment) | Expanded Extraction Station (8095-06) |
| Type | SAX Quaternary Amine Extraction Disk |
| Quantity | 20/Pk. |
| Diameter (Metric) Outer | 50 mm |