missing translation for 'onlineSavingsMsg'
Learn More

BAKERBOND spe™ Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns, J.T. Baker™

Catalog No. 02002353
Change view
Click to view available options
Quantity:
30/Box
400/Box
50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002353 30/Box
02002352 50/Box
02002354 400/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02002353 Supplier Avantor Supplier No. 709007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable Extraction Columns

Chemical Identifiers

CAS 126850-16-6

Specifications

Odor Odorless
Quantity 30/Box
Physical Form Solid
Solubility Information Practically Insoluble
Chemical Name or Material Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns