missing translation for 'onlineSavingsMsg'
Learn More

Baker-flex™, Cellulose PEI-F, J.T. Baker™

Catalog No. p-5548244
Change view
Click to view available options
Quantity:
25 Sheets
50 Sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002565 25 Sheets
02002564 50 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002565 Supplier Avantor Supplier No. 447404
Only null left

Flexible TLC Sheet

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
Physical Form Crystal or Powder
Solubility Information Insoluble in water
Quantity 25 Sheets
Odor Odorless
Chemical Name or Material Cellulose PEI-F