missing translation for 'onlineSavingsMsg'
Learn More

Baker-flex™, Cellulose, J.T. Baker™

Catalog No. 02002558
Change view
Click to view available options
Quantity:
25 Sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002558 25 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002558 Supplier Avantor Supplier No. 446804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flexible TLC Sheet

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
Solubility Information Insoluble in water
Quantity 25 Sheets
Odor Odorless
Chemical Name or Material Cellulose