missing translation for 'onlineSavingsMsg'
Learn More

Baker-flex™, Aluminum Oxide IB, J.T.Baker™

Catalog No. 02002243
Change view
Click to view available options
Quantity:
200 Sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002243 200 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002243 Supplier Avantor Supplier No. 446602
Only null left

Flexible TLC Sheet

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Quantity 200 Sheets
Solubility Information Insoluble in water
Odor Odorless
Specific Gravity 1L equals 4.00kg
Chemical Name or Material Aluminum Oxide IB