missing translation for 'onlineSavingsMsg'
Learn More

Baicalin 90.0+%, TCI America™

Catalog No. B283525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B283525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B283525G Supplier TCI America Supplier No. B283525G

Chemical Identifiers

CAS 21967-41-9
Molecular Formula C21H18O11
Molecular Weight (g/mol) 446.364
MDL Number MFCD00134418
InChI Key IKIIZLYTISPENI-ZFORQUDYSA-N
Synonym 5,6-Dihydroxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl beta-D-Glucopyranosiduronic Acid
PubChem CID 64982
ChEBI CHEBI:2981
IUPAC Name (2S,3S,4S,5R,6S)-6-(5,6-dihydroxy-4-oxo-2-phenylchromen-7-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(C(=C(C=C3O2)OC4C(C(C(C(O4)C(=O)O)O)O)O)O)O

Specifications

Melting Point 196°C
Color Yellow
Quantity 25 g
Formula Weight 446.36
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Baicalin
RTECSNumber LZ5776910
TSCA No