missing translation for 'onlineSavingsMsg'
Learn More

Interscience BagClip

Catalog No. 16948894
Change view
Click to view available options
For Use With (Equipment):
2000 to 3500 mL Bags
400 mL Bags
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16948894 400 mL Bags
16948890 2000 to 3500 mL Bags
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16948894 Supplier Interscience Supplier No. 231040*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Temporary closing clips for bags.

  • Temporary closure for bags
  • Fits all types of bags
  • Quick, waterproof closure
  • Available to fit 400 mL bags, or 2,000 to 3,500 mL bags

Specifications

Autoclavable Not Autoclavable
Type Sliding Clip
Material Plastic
For Use With (Equipment) 400 mL Bags
Color Green
Sterility Non-sterile