missing translation for 'onlineSavingsMsg'
Learn More

Bafilomycin A1

Catalog No. aaj61835mcr
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61835MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61835MCR Supplier Thermo Scientific Chemicals Supplier No. J61835MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bafilomycin A1, CAS # 88899-55-2, is an inhibitor of vacuolar ATPases from Streptomyces griseus that has antibacterial, antifungal, and antineoplastic activity. | CAS: 88899-55-2 | C35H58O9 | 622.84 g/mol

Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases.It is used as an antibacterial, antifungal, antineoplastic and an immunosuppressive. It prevents maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes and is a macrolide antibiotic and potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases
  • It is a macrolide antibiotic, and shows fungicide activity

Application

  • Bafilomycin A1 has antibacterial, antifungal, and antineoplastic activity
  • It can be immunosuppressive
  • It can prevent maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes
  • It is a potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase)

Specifications

Type Bafilomycin A1
Physical Form Oil or Film
CAS 88899-55-2
Color Colorless
Quantity 1 mg
Molecular Formula C35H58O9
MDL Number MFCD06795130
Beilstein 4730700
Solubility Information Soluble in methanol,ethanol,acetone and chloroform. Insoluble in water.
InChI Key XDHNQDDQEHDUTM-XRCNURIOSA-N
SMILES CC1CC(=CC=CC(C(OC(=O)C(=CC(=CC(C1O)C)C)OC)C(C)C(C(C)C2(CC(C(C(O2)C(C)C)C)O)O)O)OC)C
IUPAC Name (3Z,5E,7S,8R,9R,11Z,15R,16S)-16-[(2R,3S,4R)-4-[(2S,4S,5R,6S)-2,4-dihydroxy-5-methyl-6-propan-2-yloxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
Molecular Weight (g/mol) 622.84
PubChem CID 133268469
Formula Weight 622.83
Chemical Name or Material Bafilomycin A1
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber RN9781000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only