missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ M Broth

Catalog No. DF0940174
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0940174 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0940174 Supplier BD Supplier No. 294020
Edge

Used for cultivating Salmonella in foods and feeds by the accelerated enrichment serology (ES) procedure.

M Broth contains all the nutrients necessary for good growth and flagella development of Salmonella.

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Quantity 500 g
Species Salmonella species