missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Bacto™ Brain Heart Infusion

Catalog No. p-4902052
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0037178 500 g Bottle
DF0037070 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0037178 Supplier BD Difco™ Supplier No. 237500
Only null left

Used in the cultivation of fastidious and nonfastidious microorganisms. BD Difco™ Bacto™ Brain Heart Infusion is a general-purpose medium used to cultivate aerobic and anaerobic bacteria from a variety of clinical and nonclinical materials.

BHI broth is used for the cultivation of a wide variety of microorganisms, including bacteria (streptococci, pneumococci and meningococci), yeasts and molds. It serves as a base for supplemented media containing 0.1% agar, Fildes enrichment or 6.5% sodium chloride.

  • To use dehydrated product: suspend 37g in 1L of purified water
  • Displays a good cultural response to Neisseria meningitides 13090, Streptococcus pneumonia 6305 and Streptococcus pyogenes 19615
  • pH (3.7% solution, 25°C): 7.4 ± 0.2

Certifications

AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA

Specifications

Certifications/Compliance AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Light tan
Content And Storage 2 to 25°C
Form Dehydrated (free-flowing, homogenous)
Media Type Brain Heart Infusion
Product Type Nutrient Agar
Formulation Calf brain (infusion from 200g) 7.7g/L, Beef heart (infusion from 250g) 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms, including aerobic and anaerobic bateria, yeasts and molds
Packaging Type Bottle
Quantity 500 g