missing translation for 'onlineSavingsMsg'
Learn More

BD Bactec™ Media: Myco/F Lytic

Catalog No. p-4907892
Change view
Click to view available options
Quantity:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23032514 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23032514 Supplier BD Supplier No. 442288

For use with BACTEC 9240, 9120, and 9050 Systems

  • Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
  • Fluorescent series bar-coded blood culture media
  • Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
  • Vials ready to use as received—require no reconstitution or dilution
  • Detection is limited to microorganisms that will grow in a particular type of medium

Specifications

Content And Storage 2 to 25°C
Form Liquid
Media Type Myco/F Lytic
Product Type Blood Culture Medium
Formulation Processed Water 40mL, 7H9 middlebrook broth base without phosphate Salts 0.12% w/v, Brain heart infusion 0.5% w/v, Casein Hydrolysate 0.10% w/v, Supplement H 0.10% w/v, Inositol 0.05% w/v, Glycerol 0.10% w/v, Sodium Polyanetholsulfonate 0.025% w/v, Polyso
For Use With (Equipment) Bactec™ 9240, 9120 and 9050 System
Packaging Type Vial
Quantity 50/Pk.