Learn More
Sheldon Mfg Inc BACRON 600 ANAEROBIC CHAMBER DFS Item
Supplier:  Sheldon Manufacturing BACTRON600
ANAEROBIC CHAMBER 600 PETRI PLATE CAPACITY INCUBATOR, LARGE PASS BOX (12" x 12"), 115V. The BACTRON600 system permits volume handling of anaerobic cultures with maximum operator comfort. With a 17.6 cu.ft. workspace and a 600 plate capacity incubator, it is the largest, most complete anaerobic environmental system available. It is designed for high volume, yet requires far less bench space than expected for its capacity and performance features. Bactron systems have airtight construction of stainless steel and rigid Plexiglas for unobstructed vision and integrity. The cuffs form a comfortable seal around the operators arms permitting bare hand manipulation of plates and specimens inside the working chamber, making work more comfortable and efficient. In addition, all Bactron chambers have our new ergonomic armport doors, which ensure user comfort and minimize shoulder fatique. More information available at www.shellab.com
Catalog No. NC0861111
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions