missing translation for 'onlineSavingsMsg'
Learn More

BacPlaque™ Agarose, Novagen™,, MilliporeSigma™

Catalog No. 700343
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
700343 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 700343 Supplier MilliporeSigma Supplier No. 700343

For optimal plaque assays

Recommended Storage

15°C to 30°C

Specifications

Quantity 30 g
Packaging Plastic Ampoule