missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BacMam Histone H3 Reagent

Catalog No. A14154
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14154 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14154 Supplier Invitrogen™ Supplier No. A14154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BacMam Histone H3 Reagent provides a convenient genetic delivery tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest.

The BacMam Histone H3 Reagent provides a convenient genetic delivery tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest. The GFP fusion enables sensitive detection of post-translational modifications using LanthaScreen™ TR-FRET Cellular Assay technology.

The BacMam Histone H3 Reagent Lets You:

  • Use your cell background of choice with the portability of BacMam
  • Conserve precious cell samples with the miniaturizable homogenous assay format
  • Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
  • Improve data quality with the advantages of TR-FRET

Get More Physiologically Relevant Results

The BacMam Histone H3 allows the investigation of post-translational modifications of histone H3 in your choice of cellular backgrounds, including primary cells. This enables screening for potential inhibitors of histone-modifying enzymes such as kinases, acetylases, deacetylases, methyl transferases, and demethylases in their natural complexes in a physiologically relevant cell type.

BacMam and LanthaScreen™ Convenience Saves Sample and Time

When used with a LanthaScreen™ Tb-anti-Histone H3 antibody against a specific modification, assays can be run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as Western blotting and ELISA. By using the same BacMam reagent for GFP-Histone H3, a variety of histone H3 modifications (Ph, Me, Ac) can be monitored just by exchanging the Terbium (Tb) labeled antibody. In addition, the application of the LanthaScreen™ technology includes all of the advantages of TR-FRET detection, including reduced data noise, less interference from fluorescent compounds, and high sensitivity, allowing the use of fewer cells.

Specifications

Content And Storage Store the BacMam Histone H3 Reagent at 4°C
Delivery Type Baculovirus (BacMam System)
Quantity 100 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.