missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Background Plate, Fast 96-well

Catalog No. 4481308
Encompass
Change view
Click to view available options
Quantity:
1 x 96 well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4481308 1 x 96 well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4481308 Supplier Applied Biosystems™ Supplier No. 4481308
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to maintain your real-time PCR instrument with 96-well fast block

  • The Fast 96-Well Background Plate is used to maintain your real-time PCR instrument with 96-well fast block.
  • Background calibration should be run at least every six months.
  • Please see your instrument user guide for full instructions on use.

Specifications

Format 96-well Plate
Content And Storage Store at –20°C ± 5°C.

For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed Fast
Product Type Background Plate, PCR
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 96 well plate
Calibration Type Spectral Calibration
GC-Rich PCR Performance Low
PCR Method qPCR