Cells that enable the most challenging library construction and screening.Highest available transformation efficiencies for library construction and screening.Optimized for high yields of DNA.Competent Cells include Control DNA and Recovery Medium, and are packaged as DUOs (2 transformations per tube) or FIVERs (5 transformations per tube), as indicated. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.