Learn More
ACROBiosystems Inc Recombinant Protein;Mouse B7-H5 / Gi24 / VISTA Protein;HEK293;1MG;B75,B7-H5,C10orf54,Sisp-1,SISP1,Gi24,VISTA,B7H5
Catalog No. 501658419
Encompass
Recombinant Protein;Mouse B7-H5 / Gi24 / VISTA Protein;1MG;Mouse B7-H5, His Tag (B75-M52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 191 (Accession # NP_001153044.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.7kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501658419 Supplier ACROBiosystems Inc Supplier No. B75M52H71MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Mouse B7-H5 / Gi24 / VISTA Protein;1MG;Mouse B7-H5, His Tag (B75-M52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 191 (Accession # NP_001153044.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.7kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.