Recombinant Protein;200UG;Biotinylated Human B7-H5, Avitag,His Tag (B75-H82E1) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 20.8 kDa. The protein migrates as 36-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.