Learn More
Brady™ B33 Series High Adhesion Preprinted Warning Labels
Ultra Aggressive Polyester (B-483) is perfect for labeling products and general ID and can be applied to powder coated, smooth, textured or irregular surfaces.
Supplier:  Brady™ B3325483ANSIWA
 View more versions of this product
Catalog No. 17001001
Only null left
Description
Description
ToughBond™ Ultra Aggressive Polyester (B-483) is perfect for labeling products and general ID and can be applied to powder coated, smooth, textured or irregular surfaces. A ribbon is required to print this material.
  • Glossy white polyester with permanent rubber-based adhesive is thermal transfer printable
  • Material is designed for high adhesion to textured metals and low surface energy plastics
  • UL approved, CSA accepted
  • Wide service temperature range
Specifications
Chemical Identifiers
Permanent (Rubber-based)
Gloss
Polyester (B-483)
Non-sterile
Brady™ Label Printers
-40°F to 248°F
Black, Orange on White
Die-cut
Thermal Transfer
WARNING
1 Label
0.004 in.
Specifications
Permanent (Rubber-based)
Black, Orange on White
Die-cut
Single-side Printable
Non-sterile
Barcode Labeling, Equipment Labeling, Data and Telecommunications Labeling, Inventory and Inspection Labeling, General Identification
1 Label
-40°F to 248°F
4 in.
6 in.
None
Gloss
Polyester (B-483)
Thermal Transfer
WARNING
Brady™ Label Printers
250 Labels
0.004 in.
101.6 mm
Product Suggestions
Product Suggestions