missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab™ B-Plus™ Fixative
Description
- Safe and effective alternative to mercury-based fixatives
Specifications
Specifications
| Color | Blue |
| Packaging Type | Bottle |
| Quantity | 4 x 1 gal. |
| For Use With (Application) | Tissue Fixation |