missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ B-Plus™ Fixative

Catalog No. STLBFIX0507
Change view
Click to view available options
Packaging Type:
Bottle
Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
STLBFIX0507 Vial
STLBFIX1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLBFIX0507 Supplier StatLab™ Supplier No. BFIX0507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, if in contact with skin, or if inhaled.

A proprietary fixative ideal for bone marrow and lymph node biopsies.

  • Safe and effective alternative to mercury-based fixatives

Specifications

Color Blue
Packaging Type Vial
Quantity 96 x 20 mL
For Use With (Application) Tissue Fixation