missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals B-9430

$564.90 - $564.90

Chemical Identifiers

CAS 180981-09-3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66729MCR
View Documents
Thermo Scientific Chemicals
J66729MCR
1 mg
Each for $564.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

180981-09-3

Specifications

180981-09-3
Store at -20°C; Store under Nitrogen
1338.584
1 mg
White
Solid
B-9430
Air & Light Sensitive
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only