missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals B-9430

Catalog No. AAJ66729MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66729MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66729MCR Supplier Thermo Scientific Chemicals Supplier No. J66729MCR

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 180981-09-3

Specifications

Color White
Content And Storage Store at -20°C; Store under Nitrogen
Format Solid
Molecular Weight (g/mol) 1338.584
Product Type B-9430
Quantity 1 mg
Sensitivity Air & Light Sensitive

RUO – Research Use Only