missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Electrophysiology Kit

Catalog No. a1413701
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1413701 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1413701 Supplier Gibco™ Supplier No. A1413701
Only null left

B-27™ Electrophysiology Kit

The B-27™ Electrophysiology Kit contains Neurobasal™ Medium, Electro and B-27™ Supplement, Electro, both specifically developed to enhance network spike rates in cultured neurons when compared with other media and supplement combinations. Electrophysiology is the fundamental technique to assess such electrical activities to study the functions and dysfunctions in cultured neurons.

Using the B-27™ Electrophysiology Kit in your neuronal studies offers:

  • Increased synapse densities
  • More spontaneous spike rates
  • Improved multielectrode array efficiency

    More Synapses, More Signal, Simpler Testing
    The B-27™ Electrophysiology Kit is an improvement to Neurobasal™ and B-27™ combination for cultured networks. Hippocampal and cortical neurons cultured in this media system show an increased density of synapses and neurotransmitter receptors which produce higher spike rates in neural networks, helping to simplifying your study.

    Relevant Results for Your Electrophysiology Studies
    Using Invitrogen™ B-27™ Electrophysiology Kit, allows you to get more relevant results when studying neuron physiology and the biochemical basis of normal and disease processes. The B-27™ Electrophysiology Kit can yield better results in other electrophysiological studies, including neurons used in multielectrode arrays in neurotoxicology and neuropharmacology studies.

    For other applications, we offer alternative formulations of B-27™ and Neurobasal™.
  • Specifications

    Content And Storage B-27 Supplement, Electro (A14097-01)
    Store at -5°C to -20°C

    Neurobasal Media, Electro (A14098-01)
    Store at 2°C to 8°C
    Cell Type Neural Cells
    Classification Serum-free
    Culture Type Mammalian Cell Culture
    Form Liquid
    Product Type Electrophysiology Kit
    Serum Level Serum-Free
    Sterility Sterile
    Product Line Gibco
    Quantity 1 Kit

    For Research Use Only. Not for use in diagnostic procedures.