missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems AzureSpectra Fluorescent Western Blot Kit with Protein-Free Block
Kit contains materials for 10 fluorescent Western blots, including LF PVDF membranes, wash buffer, protein-free blocking buffer, and secondary antibodies.
Supplier: Azure Biosystems AC2140
Description
- Full kits for two-color fluorescent Western blots. Also offered with Fluorescent Block.
- Each kit contains enough materials for 10 Western blots:
- 9 x 7 cm Low Fluorescence PVDF Membranes, 10 membranes
- 10 x Fluorescent Blot Washing Buffer, 250 mL
- 5 x Protein-Free Blot Blocking Buffer, 60 mL
- 2 x Background Quenching Sheets
- 2 x 40 μL Fluorescent Secondary Antibodies
Specifications
| Western Blot Kit | |
| Western Blot | |
| 10 Mini Blots | |
| PVDF Membranes, Wash Buffer, Protein-free Blocking Buffer, Secondary Antibodies rb700/ms800 |
| Fluorescence | |
| Nitrocellulose, PVDF | |
| Includes: Secondary Ab rb700/ms800 |