missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems AzureSpectra Fluorescent Western Blot Kit with Protein-Free Block
Description
- Full kits for two-color fluorescent Western blots. Also offered with Fluorescent Block.
- Each kit contains enough materials for 10 Western blots:
- 9 x 7 cm Low Fluorescence PVDF Membranes, 10 membranes
- 10 x Fluorescent Blot Washing Buffer, 250 mL
- 5 x Protein-Free Blot Blocking Buffer, 60 mL
- 2 x Background Quenching Sheets
- 2 x 40 μL Fluorescent Secondary Antibodies
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | Secondary Antibodies rb650/ms550 |
| Quantity | 10 Mini Blots |
| Kit Contents | Includes: Secondary Ab rb650/ms550 |
| Content And Storage | PVDF Membranes, Wash Buffer, Protein-free Blocking Buffer, Secondary Antibodies rb650/ms550 |