missing translation for 'onlineSavingsMsg'
Learn More

Azure C

CAS: 531-57-7 | C13H12ClN3S | 277.77 g/mol

Supplier:  Thermo Scientific Chemicals J6451503

 View more versions of this product

Catalog No. AAJ6451503

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Azure C
C13H12ClN3S
4060661
C.I. 52002; 3-Amino-7-(methylamino)phenothiazin-5-ium chloride
C[NH+]=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1.[Cl-]
277.77
277.78
1 g
531-57-7
MFCD00067677
14,929
DDGMDTGNGDOUPX-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)-methylazanium;chloride
68277
Black or Green
Powder

Chemical Identifiers

531-57-7
277.77
DDGMDTGNGDOUPX-UHFFFAOYSA-N
68277
C[NH+]=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1.[Cl-]
C13H12ClN3S
MFCD00067677
C.I. 52002; 3-Amino-7-(methylamino)phenothiazin-5-ium chloride
(7-aminophenothiazin-3-ylidene)-methylazanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-512-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only