missing translation for 'onlineSavingsMsg'
Learn More

Azure C

Catalog No. AAJ6451506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6451506 5 g
AAJ6451503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6451506 Supplier Thermo Scientific Chemicals Supplier No. J6451506

CAS: 531-57-7 | C13H12ClN3S | 277.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 531-57-7
Molecular Formula C13H12ClN3S
Molecular Weight (g/mol) 277.77
MDL Number MFCD00067677
InChI Key DDGMDTGNGDOUPX-UHFFFAOYSA-N
Synonym C.I. 52002; 3-Amino-7-(methylamino)phenothiazin-5-ium chloride
PubChem CID 68277
IUPAC Name (7-aminophenothiazin-3-ylidene)-methylazanium;chloride
SMILES C[NH+]=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1.[Cl-]

Specifications

Beilstein 4060661
Merck Index 14,929
Color Black or Green
Quantity 5 g
Physical Form Powder
Formula Weight 277.78
Chemical Name or Material Azure C
EINECSNumber 208-512-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only