missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure B Certified, MP Biomedicals™
Description
- Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues.
- Useful as a stain for Negri Bodies.
- Can be used in the Romanowsky-type stains for blood and blood protozoa.
Specifications
Specifications
| InChI Key | KFZNPGQYVZZSNV-UHFFFAOYSA-M |
| SMILES | [Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C |
| IUPAC Name | 3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride |
| Molecular Weight (g/mol) | 305.82 |
| Grade | Certified |
| Color Index No. | 52010 |
| Quantity | 1 g |
| Physical Form | Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.