Learn More
Azure A
For staining cell granules and tissues | CAS: 531-53-3 | C14H14ClN3S | 291.797 g/mol
Supplier: Thermo Scientific Chemicals J6134622
Description
For staining cell granules and tissues
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Azure A | |
| 531-53-3 | |
| MFCD00012112 | |
| 14,927 | |
| NALREUIWICQLPS-UHFFFAOYSA-N | |
| (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride | |
| 13735 | |
| Green | |
| Powder |
| 290°C (decomposition) | |
| C14H14ClN3S | |
| 3922287 | |
| C.I. 52005 | |
| C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-] | |
| 291.797 | |
| 291.8 | |
| 100 g |
Chemical Identifiers
| 531-53-3 | |
| 291.797 | |
| NALREUIWICQLPS-UHFFFAOYSA-N | |
| 13735 | |
| C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-] |
| C14H14ClN3S | |
| MFCD00012112 | |
| C.I. 52005 | |
| (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride |
Safety and Handling
EINECSNumber : 208-510-7
RTECSNumber : SP5660000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only