missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Azur eosin methylene-blue, pure, solution according to Giemsa

Catalog No. AC295595000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC295595000 500 mL
AC295591000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295595000 Supplier Thermo Scientific Chemicals Supplier No. 295595000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51811-82-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Azur eosin methylene-blue
CAS 51811-82-6, 67-56-1
Lambda Max 523 to 526nm (c=0.1, MeOH) (eosine), 646 to 651nm (c=0.1, MeOH) (methylene blue)
Packaging Glass bottle
MDL Number MFCD00081642
Color Blue
Quantity 500 mL
Physical Form Solution
Grade Pure

RUO – Research Use Only