missing translation for 'onlineSavingsMsg'
Learn More

Azulene 99.0+%, TCI America™

Catalog No. p-7116979
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0634100MG 100 mg
A06341G 1 g
A06345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0634100MG Supplier TCI America Supplier No. A0634100MG
Only null left

Chemical Identifiers

CAS 275-51-4
Molecular Formula C10H8
Molecular Weight (g/mol) 128.174
MDL Number MFCD00003810
InChI Key CUFNKYGDVFVPHO-UHFFFAOYSA-N
Synonym cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep
PubChem CID 9231
ChEBI CHEBI:31249
IUPAC Name azulene
SMILES C1=CC=C2C=CC=C2C=C1

Specifications

Melting Point 100°C
Color Purple-Red
Boiling Point 242°C
Quantity 100 mg
Formula Weight 128.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Azulene
RTECSNumber CO4570000
TSCA Yes