missing translation for 'onlineSavingsMsg'
Learn More

Azulene, 99%

Catalog No. AAL08271MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08271MD 250 mg
AAL0827103 1 g
AAL0827106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL08271MD Supplier Thermo Scientific Chemicals Supplier No. L08271MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 275-51-4 | C10H8 | 128.174 g/mol

Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 275-51-4
Molecular Formula C10H8
Molecular Weight (g/mol) 128.174
MDL Number MFCD00003810
InChI Key CUFNKYGDVFVPHO-UHFFFAOYSA-N
Synonym cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep
PubChem CID 9231
ChEBI CHEBI:31249
IUPAC Name azulene
SMILES C1=CC=C2C=CC=C2C=C1

Specifications

Melting Point 98°C to 101°C
Boiling Point 241°C to 243°C
Quantity 250 mg
Beilstein 969517
Merck Index 14,926
Solubility Information Soluble in organic solvents.
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material Azulene
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-993-6
RTECSNumber CO4570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only