missing translation for 'onlineSavingsMsg'
Learn More

Azoxybenzene, 98+%, Thermo Scientific Chemicals

$81.56 - $81.56

Chemical Identifiers

CAS 495-48-7
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00019925
InChI Key GAUZCKBSTZFWCT-UHFFFAOYSA-N
Synonym azoxybenzene, azoxybenzide, azoxybenzol, fenazox, fentoxan, azobenzene, oxide, diphenyldiazene 1-oxide, benzene, azoxydi, diazene, diphenyl-, 1-oxide, ordinary azoxybenzene
PubChem CID 10316
ChEBI CHEBI:51865
IUPAC Name oxido-phenyl-phenyliminoazanium
SMILES C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2002109
View Documents
Thermo Scientific Chemicals
B2002109
10 g
Each for $81.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

495-48-7
198.225
GAUZCKBSTZFWCT-UHFFFAOYSA-N
10316
oxido-phenyl-phenyliminoazanium
C12H10N2O
MFCD00019925
azoxybenzene, azoxybenzide, azoxybenzol, fenazox, fentoxan, azobenzene, oxide, diphenyldiazene 1-oxide, benzene, azoxydi, diazene, diphenyl-, 1-oxide, ordinary azoxybenzene
CHEBI:51865
C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]

Specifications

495-48-7
1.16
C12H10N2O
10 g
14,923
GAUZCKBSTZFWCT-UHFFFAOYSA-N
oxido-phenyl-phenyliminoazanium
10316
198.23
Azoxybenzene
33°C to 36°C
130°C (0.9 mmHg)
MFCD00019925
743984
azoxybenzene, azoxybenzide, azoxybenzol, fenazox, fentoxan, azobenzene, oxide, diphenyldiazene 1-oxide, benzene, azoxydi, diazene, diphenyl-, 1-oxide, ordinary azoxybenzene
C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]
198.225
CHEBI:51865
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 207-802-1

RTECSNumber : CO4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only