missing translation for 'onlineSavingsMsg'
Learn More

Azomethine H 97.0+%, TCI America™

Catalog No. p-7118260
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50165G 5 g
A501625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50165G Supplier TCI America Supplier No. A50165G
Only null left

Chemical Identifiers

CAS 5941-07-1
Molecular Formula C17H14NNaO9S2
Molecular Weight (g/mol) 463.41
MDL Number MFCD00042050
InChI Key BEDIAJJRAYEWBI-TTWKNDKESA-M
Synonym 4-hydroxy-5-salicylideneamino-2,7-naphthalenedisulfonic acid sodium salt
PubChem CID 123134089
IUPAC Name 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na]

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 445.39
Percent Purity ≥97.0% (T)
Chemical Name or Material Azomethine H [Spectrophotometric Reagent for B]