missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azomethine H, MP Biomedicals™
Chemical Identifiers
| CAS | 5941-07-1 |
|---|---|
| Molecular Formula | C17H14NNaO9S2 |
| Molecular Weight (g/mol) | 463.41 |
| MDL Number | MFCD00149588 |
| InChI Key | BEDIAJJRAYEWBI-TTWKNDKESA-M |
| Synonym | 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15416910
|
N/A | 10 g |
N/A
|
|
|||||
Chemical Identifiers
| 5941-07-1 | |
| 463.41 | |
| BEDIAJJRAYEWBI-TTWKNDKESA-M |
| C17H14NNaO9S2 | |
| MFCD00149588 | |
| 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate |
Specifications
| 5941-07-1 | |
| Extinction Coefficient max: 237nm C=0.01% ≥50, 000 (Lit.) | |
| MFCD00149588 | |
| BEDIAJJRAYEWBI-TTWKNDKESA-M | |
| 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium | |
| 123134089 | |
| Orange | |
| Azomethine H, Chemistry |
| 1.04 | |
| C17H14NNaO9S2 | |
| 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate | |
| C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na] | |
| 463.41 | |
| 423.4 | |
| Powder |
Safety and Handling
EINECSNumber : 227-698-1
Recommended Storage : Store at 4°C.