missing translation for 'onlineSavingsMsg'
Learn More

Azomethine H monosodium salt monohydrate, 95%

Catalog No. p-7077657
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457090050 5 g
AC457090250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457090050 Supplier Thermo Scientific Chemicals Supplier No. 457090050
Only null left

CAS: 304655-82-1 | C17H15NNaO9S2 | 464.415 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 304655-82-1
Molecular Formula C17H15NNaO9S2
Molecular Weight (g/mol) 464.415
InChI Key CYPHYAYBFNUBEV-TTWKNDKESA-N
PubChem CID 131877441
IUPAC Name 4-hydroxy-5-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium;hydrate
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.O.[Na]

Specifications

Infrared Spectrum Authentic
Formula Weight 463.41
Percent Purity 95%
Quantity 5 g
Chemical Name or Material Azomethine H monosodium salt monohydrate
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only