missing translation for 'onlineSavingsMsg'
Learn More

Azomethine-H monosodium salt hydrate

Catalog No. AAA1092706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1092706 5 g
AAA1092703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1092706 Supplier Thermo Scientific Chemicals Supplier No. A1092706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 206752-32-1 | C17H12NNaO8S2 | 445.39 g/mol

Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

Solubility
Soluble in water, ethanol and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.

Chemical Identifiers

CAS 206752-32-1
Molecular Formula C17H12NNaO8S2
Molecular Weight (g/mol) 445.39
MDL Number MFCD00010498
InChI Key GRYBZNLNHVSHPY-UHFFFAOYSA-M
Synonym sodium 5-hydroxy-4-2-hydroxybenzylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, sodium 5-hydroxy-4-e-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, azomethin-h monosodium salt hydrate, 4-hydroxy-5-e-2-hydroxyphenyl methylidene amino-7-sodiooxysulfonyl naphthalene-2-sulfonic acid hydrate, sodium 5-hydroxy-4-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate
PubChem CID 122198137
SMILES [Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4100175
Solubility Information Soluble in water,ethanol and acetone.
IUPAC Name sodium;4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]-7-sulfonaphthalene-2-sulfonate;hydrate
Formula Weight 445.41 (Anhydrous)
Chemical Name or Material Azomethine-H monosodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 250-975-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only