missing translation for 'onlineSavingsMsg'
Learn More

Azoic Diazo Component 20 (Base) 95.0+%, TCI America™

Catalog No. a198625g
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
A198625G 25 g
1 options

Catalog No. A198625G

Supplier: TCI America A198625G

Only null left

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym fast blue bb, fast blue bb base, n-4-amino-2,5-diethoxyphenyl benzamide, 4'-amino-2',5'-diethoxybenzanilide, blue 2b base, blue salt nbb, fast blue bbn, stabamine blue bb, fast blue base bb, fast blue eb base
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2

Specifications

Melting Point 101°C
Color Gray-Red
Quantity 25 g
Formula Weight 300.36
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Azoic Diazo Component 20 (Base)
EINECSNumber (5)-2311
TSCA Yes