missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene-4,4'-dicarboxylic Acid 95.0+%, TCI America™

Catalog No. p-7117542
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15961G 1 g
A15965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15961G Supplier TCI America Supplier No. A15961G

Chemical Identifiers

CAS 586-91-4
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD00496621
InChI Key NWHZQELJCLSKNV-UHFFFAOYSA-N
Synonym 4,4′C-Azodibenzoic Acid
PubChem CID 11471
IUPAC Name 4-[(4-carboxyphenyl)diazenyl]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N=NC2=CC=C(C=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 270.24
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene-4,4′-dicarboxylic Acid