missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene-4,4'-dicarbonyl Dichloride 98.0+%, TCI America™

Catalog No. A15751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15751G 1 g
A15755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15751G Supplier TCI America Supplier No. A15751G
Only null left

Chemical Identifiers

CAS 10252-29-6
Molecular Formula C14H8Cl2N2O2
Molecular Weight (g/mol) 307.13
MDL Number MFCD00053223
InChI Key ASOXKYGOZZTVHL-UHFFFAOYSA-N
PubChem CID 4246116
IUPAC Name 4-[(4-carbonochloridoylphenyl)diazenyl]benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)N=NC2=CC=C(C=C2)C(=O)Cl

Specifications

Melting Point 164°C
Quantity 1 g
UN Number 3261
Synonym 4,4′C-Azodibenzoyl Dichloride
Formula Weight 307.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene-4,4′-dicarbonyl Dichloride