missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Azilsartan

Catalog No. AAH3701803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3701803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3701803 Supplier Thermo Scientific™ Supplier No. H37018MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 147403-03-0
Melting Point 188°C dec.
Molecular Formula C25H20N4O5
Quantity 1 g
MDL Number MFCD00916395
InChI Key KGSXMPPBFPAXLY-UHFFFAOYSA-N
SMILES CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1
IUPAC Name 2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
Molecular Weight (g/mol) 456.46
Formula Weight 456.46
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 808-058-6