missing translation for 'onlineSavingsMsg'
Learn More

Azidotrimethylsilane, 94%

Catalog No. AC167930100
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167930100 10 mL Glass bottle
AC167930500 50 mL Glass bottle
AC167932500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167930100 Supplier Thermo Scientific Chemicals Supplier No. 167930100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
IUPAC Name azido(trimethyl)silane
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Color Colorless to Yellow
Density 0.8760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4142 to 1.4162
Linear Formula (CH3)3SiN3
Quantity 10 mL
Fieser 01,1236; 03,316; 05,719; 06,632; 07,394; 09,21; 10,14; 11,32; 12,37; 13,24; 14,25; 15,16; 16,17; 17,378
Specific Gravity 0.876
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, dichloromethane and benzene
Formula Weight 115.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Azidotrimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 225-078-5

RUO – Research Use Only