missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-maleimide Kit

Catalog No. aaj65984mb
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65984MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65984MB Supplier Thermo Scientific Chemicals Supplier No. J65984MB
Only null left

C15H23N5O6

An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

Solubility
Difficult to mix.

Notes
Kit contains two components: a) 1.1 eq. of azido-PEG3-amine as an oil; b) 1.0 eq. of maleimide-NHS ester as a solid.

Chemical Identifiers

Molecular Formula C15H23N5O6
MDL Number MFCD22380764
Synonym N-(11-Azido-3,6,9-trioxaundecyl)-3-(N-maleimidyl)propionamide

Specifications

Quantity 25 mg
UN Number UN2735
Solubility Information Difficult to mix.
Formula Weight 369.37
Chemical Name or Material Azido-PEG3-maleimide Kit
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P305+P351+P338-P302+P352-P321-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only