missing translation for 'onlineSavingsMsg'
Learn More

Azetidine Hydrochloride 95.0+%, TCI America™

Catalog No. A24511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24511G 1 g
A24515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24511G Supplier TCI America Supplier No. A24511G

Chemical Identifiers

CAS 36520-39-5
Molecular Formula C3H8ClN
Molecular Weight (g/mol) 93.554
MDL Number MFCD00191762
InChI Key HGQULGDOROIPJN-UHFFFAOYSA-N
Synonym azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
PubChem CID 12308726
IUPAC Name azetidine;hydrochloride
SMILES C1CNC1.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 93.55
Percent Purity ≥95.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Azetidine Hydrochloride